ContactUsLinkComponent
Elbow stopcock, b.safe
Elbow stopcock, b.safe
Catalog # BOHLR1201-05
Supplier:  Bohlender
Elbow stopcock, b.safe
Catalog # BOHLR1201-05
Supplier:  Bohlender
Specifications
  • Hole Ø:
    10 mm
  • Thread:
    GL 25 (Male and female)
  • Pk:
    1
Specifications
About this item
90° Elbow fitting. To connect b.safe Waste Tubes to b.safe Waste Caps without bending. Integrated stopcock with plug made of PTFE-EX to stop the flow through b.safe Waste Tubes when canisters are to be exchanged or emptied.
  • Made of static dissipative PE-EX
  • Firmly mounted union nut made of PPS-EX with female thread GL 25
  • Outer dimension: 34×52×102 mm
To connect b.safe Waste Tubes to b.safe Waste Caps GL 25 without bending. No dripping from b.safe Waste Tubes during exchange of collection canisters.