Über diesen Artikel
These permanent labels are designed for the identification of tubes, vials, boxes, and other plastic or cardboard containers stored in ultra-low temperature laboratory freezers.
- Temperature range: –80 to +110 °C (–112 to +230 °F)
- Ideal for long-term storage in deep freeze conditions (–80 °C)
- No cure time; can be stored in freezers immediately after application
- No additional lamination necessary
- Labels and printout resist spraying with alcohols and other chemicals
- Printable with all major thermal-transfer printer brands
- Available in a large range of sizes, colours, and configurations
FJT barcode labels are perfect for identifying small diameter vials, microtubes, Falcon tubes, cell culture plates, microplates, and polypropylene and cardboard freezer boxes. These freezer-grade labels are waterproof and tear-proof and will adhere to containers in ultra-low temperature (–80 °C/–112 °F) lab freezers and dry ice without the need for extra lamination.
Available in multiple configurations and sizes, with a permanent adhesive. They are made of a durable thermoplastic film and will resist spraying with alcohols, bleach, detergents, as well as wiping with surface cleaning wipes.