Spécifications
- With:8× UNF ¼" capillary connectors, 4× NPT ¹/₈" hose connectors
- Environmentally Preferable:
- Pk:1 SET
Spécifications
À propos de cet article
One connection for collecting liquid waste from up to 4 HPLC systems that can be integrated directly in laboratory hoods or working tables. b.safe Waste Tubes lead liquid solvent waste directly into connected canisters.
- Compact collective connector to combine the outlets of two respectively up to four HPLC units
- All connectors are placed sideways
- Additional slope improves the flow and prevents any backflow
Made of static dissipative PE-EX, suitable for boards with a thickness of 10 to 35 mm, easy fixing with counter nut. Low installation height due to connections at the sides, inclined threads are for additional sloping and connection of tubings without bending.
Connections at upper side: 4×2 capillary connections (thread UNF ¼") incl. b.safe Fittings to connect capillaries with ext.Ø 1,6/2,2/3,2 mm , 4×1 hose connection (thread NPT ¹/₈") incl. b.safe Hose Connector for tubing Int.Ø 6,5 to 8 mm. Connection at bottom: male thread GL 25 for direct connection with flexible b.safe Waste Tubes.
1× b.safe Desktop Mount Multiport IV
24× b.safe Fittings: 8× Ø 1,6 mm (green), 8× Ø 2,2 mm (purple), 8× Ø 3,2 mm (yellow)
8× b.safe Blind Fittings for capillary connection (UNF ¼")
4× b.safe Hose Connectors for Int.Ø 6,5 to 8 mm
4× b.safe Blind Fittings for hose connection (NPT ¹/₈")