Orders LinkContactUsLinkComponent
ISO warning labels
ISO warning labels
Supplier:   GA
ISO warning labels
Supplier:  GA
Certificates
ISO warning labels
ISO warning labels
3 Options Available Below
Product Details & Documents
Pre-printed general warning labels for the identification of equipment, containers, and material that may pose a safety risk in the laboratory environment.
  • Withstand high-temperature conditions up to 100° C (212° F)
  • Pre-printed with full color, yellow on silver background, design
  • Durable thermoplastic material is resistant to wear and tear, water, mild chemicals, and detergents
  • Resistant to gamma irradiation (tested to up to 50kGy)
Coated with a glove-friendly adhesive, these waterproof thermoplastic labels are perfect for labeling water baths, heat blocks, autoclave containers, and instruments or tools with moving parts.
Product Family Options
Product Information
  • Colour
    Dimensions
    Packed
    Availability
    Price
  • GAINWL-008-0.25R
    ISO warning label - Hot Surface Hazard
    ISO warning label - Hot Surface Hazard
    Black/yellow
    19,05×19,05 mm
    250 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 55.70
    SFr. 55.70
    1 * 1 ROU
       SFr. 55.70
  • GAINWL-010-0.25R
    ISO warning label - Low Temperature
    ISO warning label - Low Temperature
    Black/yellow
    19,05×19,05 mm
    250 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 55.70
    SFr. 55.70
    1 * 1 ROU
       SFr. 55.70
  • GAINWL-009-0.25R
    ISO warning label - General Warning Symbol
    ISO warning label - General Warning Symbol
    Black/yellow
    19,05×19,05 mm
    250 labels/roll
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 55.70
    SFr. 55.70
    1 * 1 ROU
       SFr. 55.70