Transparent cryogenic labels for thermal-transfer printers
: GA
Transparent thermoplastic cryogenic labels for bio-medical research and clinical laboratories.
- Labels can withstand a wide temperature range: extreme low (–196 °C) to elevated (100 °C)
- Transparent design ensures contents of labeled containers remain visible
- Printable with all major thermal-transfer printer brands
Labels will withstand long-term storage in deep-freeze and harsh laboratory environments, including liquid and vapor phase liquid nitrogen at –196 °C (–320.8 °F), –80 °C (–112 °F) freezers, and dry ice transportation.
Frequently Bought Together