Removable cryogenic labels for laser printers
Catalog #
Product Details & Documents
Cryogenic thermoplastic laser labels with a glove-friendly removable adhesive.
- Glove-friendly adhesive ensures the labels can be removed easily without leaving any glue residue
- Resist extreme cryogenic temperatures (–196 °C in the liquid phase and vapor phase liquid nitrogen)
- Printable by all major desktop laser printers
- Resistant to gamma irradiation (tested to up to 50 kGy)
Cryogenic thermoplastic laser labels are ideal for long-term storage of samples in liquid nitrogen and harsh deep-freeze environments used in biomedical research and clinical laboratories. Cryo labels accept desktop laser printing and permanent marker. Water-resistant, and wear and tear proof. Labels do not jam printers. Resistant to UV and gamma irradiation.