ESD safety shoes, lace-up, jo_Slim Low, Jori
Supplier: ELTEN
Certificates
About this item
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
- Padded tongue
- Rubber/EVA/EVA sole jo_SAMBA
- Steel toe cap
Suede/hydrophobised textile material construction with fabric lining for natural ventilation. Full-length inlay ESD sole. Non-metallic, penetration-resistant midsole.
Certifications: EN ISO 20345 S3 SRC, Form A.
Specifications
- Catalog No:
- ELTN12851/36
- ELTN12851/37
- ELTN12851/38
- ELTN12831/42
- ELTN12841/41
- ELTN12841/42
- ELTN12841/43
- ELTN12841/44
- ELTN12831/36
- ELTN12841/45
- ELTN12831/37
- ELTN12841/46
- ELTN12841/47
- ELTN12831/38
- ELTN12851/46
- ELTN12841/48
- ELTN12831/39
- ELTN12851/47
- ELTN12831/40
- ELTN12851/48
- ELTN12831/41
- ELTN12841/39
- ELTN12841/40
- ELTN12851/39
- ELTN12831/43
- ELTN12851/40
- ELTN12831/44
- ELTN12851/41
- ELTN12831/45
- ELTN12831/46
- ELTN12851/42
- ELTN12851/43
- ELTN12831/47
- ELTN12831/48
- ELTN12851/44
- ELTN12841/36
- ELTN12851/45
- ELTN12841/37
- ELTN12841/38
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001