ESD safety ankle boots, BOA® Fit System, HANNAH XXTL BOA® Mid
Nome del fornitore: ELTEN
Certificati
Informazioni su questo articolo
Ladies' safety ankle boot is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
- Full, padded bellows tongue
- PU/PU sole, WELLMAXX TRAINERS LADY
- Steel toe cap
- BOA® Fit System
- Leather-free
Hydrophobised microfibre/hydrophobised textile material construction with fabric lining for natural ventilation. Full-length inlay sole, ESD PRO Lady Y. Non-metallic, penetration-resistant midsole. Sole core made of Infinergy® by BASF.
Certifications: EN ISO 20345 S3 SRC, form B.
Specifiche
- Catalog No:
- ELTN746561/38
- ELTN746561/39
- ELTN746561/36
- ELTN746561/37
- ELTN746561/42
- ELTN746561/40
- ELTN746561/41
- ELTN746561/35
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
Specifiche
Frequently Bought Together