Orders LinkContactUsLinkComponent
Cryogenic Tapes for Laboratory Use
Cryogenic Tapes for Laboratory Use
Supplier:   GA
Cryogenic Tapes for Laboratory Use
Supplier:  GA
Certificates
Cryogenic Tapes for Laboratory Use
Cryogenic Tapes for Laboratory Use
48 Options Available Below
Product Details & Documents
Continuous writable tape for cryogenic use. Provided with a strong permanent adhesive, this cryo tape will stay firmly attached during prolonged exposure to low temperature conditions, as well as repeated freeze-thaw cycles. Available in assorted colours.
  • Temperature range: −196 to +110 °C
  • Designed for use in harsh cryogenic conditions
  • Deep-freeze permanent adhesive ensures the tape won't peel or crack when exposed to liquid nitrogen
  • Continuous tape format allows use of as much or little as necessary
Waterproof, resistant to liquid nitrogen, dry ice, multiple freeze-thaw cycles, gamma irradiation, and other harsh laboratory environments. Extra-strong permanent adhesive. Available in a variety of colours for freezer and liquid nitrogen use (as low as −196 °C) on plastic and glass containers.
Product Family Options
Product Information
  • Colour
    Length
    Width
    Availability
    Price
  • GAINTJTA-13C1-50BL
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Blue
    15 m
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 35.00
    SFr. 35.00
    1 * 1 ROU
       SFr. 35.00
  • GAINTJTA-13C1-50GA
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Green
    15 m
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 35.00
    SFr. 35.00
    1 * 1 ROU
       SFr. 35.00
  • GAINTJTA-13C1-50OR
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Orange
    15 m
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 35.00
    SFr. 35.00
    1 * 1 ROU
       SFr. 35.00
  • GAINTJTA-13C1-50RE
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Red
    15 m
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 35.00
    SFr. 35.00
    1 * 1 ROU
       SFr. 35.00
  • GAINTJTA-13C1-50WH
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    White
    15 m
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 35.00
    SFr. 35.00
    1 * 1 ROU
       SFr. 35.00
  • GAINTJTA-13C1-50YE
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Yellow
    15 m
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 35.00
    SFr. 35.00
    1 * 1 ROU
       SFr. 35.00
  • GAINTJTA13C3-100BL
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Blue
    30 m
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 70.00
    SFr. 70.00
    1 * 1 ROU
       SFr. 70.00
  • GAINTJTA13C3-100GA
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Green
    30 m
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 70.00
    SFr. 70.00
    1 * 1 ROU
       SFr. 70.00
  • GAINTJTA13C3-100OR
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Orange
    30 m
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 70.00
    SFr. 70.00
    1 * 1 ROU
       SFr. 70.00
  • GAINTJTA13C3-100RE
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Red
    30 m
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 70.00
    SFr. 70.00
    1 * 1 ROU
       SFr. 70.00
  • GAINTJTA13C3-100WH
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    White
    30 m
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 70.00
    SFr. 70.00
    1 * 1 ROU
       SFr. 70.00
  • GAINTJTA13C3-100YE
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Yellow
    30 m
    12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 70.00
    SFr. 70.00
    1 * 1 ROU
       SFr. 70.00
  • GAINTJTA-19C1-50BL
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Blue
    15 m
    19 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 36.80
    SFr. 36.80
    1 * 1 ROU
       SFr. 36.80
  • GAINTJTA-19C1-50GA
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Green
    15 m
    19 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 36.80
    SFr. 36.80
    1 * 1 ROU
       SFr. 36.80
  • GAINTJTA-19C1-50OR
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Orange
    15 m
    19 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 36.80
    SFr. 36.80
    1 * 1 ROU
       SFr. 36.80
  • GAINTJTA-19C1-50RE
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Red
    15 m
    19 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 36.80
    SFr. 36.80
    1 * 1 ROU
       SFr. 36.80
  • GAINTJTA-19C1-50WH
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    White
    15 m
    19 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 36.80
    SFr. 36.80
    1 * 1 ROU
       SFr. 36.80
  • GAINTJTA-19C1-50YE
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Yellow
    15 m
    19 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 36.80
    SFr. 36.80
    1 * 1 ROU
       SFr. 36.80
  • GAINTJTA19C3-100BL
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Blue
    30 m
    19 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 73.40
    SFr. 73.40
    1 * 1 ROU
       SFr. 73.40
  • GAINTJTA19C3-100GA
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Green
    30 m
    19 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 73.40
    SFr. 73.40
    1 * 1 ROU
       SFr. 73.40
  • GAINTJTA19C3-100OR
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Orange
    30 m
    19 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 73.40
    SFr. 73.40
    1 * 1 ROU
       SFr. 73.40
  • GAINTJTA19C3-100RE
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Red
    30 m
    19 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 73.40
    SFr. 73.40
    1 * 1 ROU
       SFr. 73.40
  • GAINTJTA19C3-100WH
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    White
    30 m
    19 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 73.40
    SFr. 73.40
    1 * 1 ROU
       SFr. 73.40
  • GAINTJTA19C3-100YE
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Yellow
    30 m
    19 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 73.40
    SFr. 73.40
    1 * 1 ROU
       SFr. 73.40
  • GAINTJTA-25C1-50BL
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Blue
    15 m
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 39.80
    SFr. 39.80
    1 * 1 ROU
       SFr. 39.80
  • GAINTJTA-25C1-50GA
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Green
    15 m
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 39.80
    SFr. 39.80
    1 * 1 ROU
       SFr. 39.80
  • GAINTJTA-25C1-50OR
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Orange
    15 m
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 39.80
    SFr. 39.80
    1 * 1 ROU
       SFr. 39.80
  • GAINTJTA-25C1-50RE
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Red
    15 m
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 39.80
    SFr. 39.80
    1 * 1 ROU
       SFr. 39.80
  • GAINTJTA-25C1-50WH
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    White
    15 m
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 39.80
    SFr. 39.80
    1 * 1 ROU
       SFr. 39.80
  • GAINTJTA-25C1-50YE
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Yellow
    15 m
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 39.80
    SFr. 39.80
    1 * 1 ROU
       SFr. 39.80
  • GAINTJTA25C3-100BL
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Blue
    30 m
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 79.60
    SFr. 79.60
    1 * 1 ROU
       SFr. 79.60
  • GAINTJTA25C3-100GA
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Green
    30 m
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 79.60
    SFr. 79.60
    1 * 1 ROU
       SFr. 79.60
  • GAINTJTA25C3-100OR
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Orange
    30 m
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 79.60
    SFr. 79.60
    1 * 1 ROU
       SFr. 79.60
  • GAINTJTA25C3-100RE
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Red
    30 m
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 79.60
    SFr. 79.60
    1 * 1 ROU
       SFr. 79.60
  • GAINTJTA25C3-100WH
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    White
    30 m
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 79.60
    SFr. 79.60
    1 * 1 ROU
       SFr. 79.60
  • GAINTJTA25C3-100YE
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Yellow
    30 m
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 79.60
    SFr. 79.60
    1 * 1 ROU
       SFr. 79.60
  • GAINTJTA-70C1-50BL
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Blue
    15 m
    69,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 110.00
    SFr. 110.00
    1 * 1 ROU
       SFr. 110.00
  • GAINTJTA-70C1-50GA
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Green
    15 m
    69,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 110.00
    SFr. 110.00
    1 * 1 ROU
       SFr. 110.00
  • GAINTJTA-70C1-50OR
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Orange
    15 m
    69,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 110.00
    SFr. 110.00
    1 * 1 ROU
       SFr. 110.00
  • GAINTJTA-70C1-50RE
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Red
    15 m
    69,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 110.00
    SFr. 110.00
    1 * 1 ROU
       SFr. 110.00
  • GAINTJTA-70C1-50WH
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    White
    15 m
    69,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 110.00
    SFr. 110.00
    1 * 1 ROU
       SFr. 110.00
  • GAINTJTA-70C1-50YE
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Cryogenic tape for laboratory use, core size: 25,4 mm
    Yellow
    15 m
    69,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 110.00
    SFr. 110.00
    1 * 1 ROU
       SFr. 110.00
  • GAINTJTA70C3-100BL
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Blue
    30 m
    69,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 219.00
    SFr. 219.00
    1 * 1 ROU
       SFr. 219.00
  • GAINTJTA70C3-100GA
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Green
    30 m
    69,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 219.00
    SFr. 219.00
    1 * 1 ROU
       SFr. 219.00
  • GAINTJTA70C3-100OR
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Orange
    30 m
    69,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 219.00
    SFr. 219.00
    1 * 1 ROU
       SFr. 219.00
  • GAINTJTA70C3-100RE
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Red
    30 m
    69,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 219.00
    SFr. 219.00
    1 * 1 ROU
       SFr. 219.00
  • GAINTJTA70C3-100WH
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    White
    30 m
    69,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 219.00
    SFr. 219.00
    1 * 1 ROU
       SFr. 219.00
  • GAINTJTA70C3-100YE
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Cryogenic tape for laboratory use, core size: 76,2 mm
    Yellow
    30 m
    69,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 219.00
    SFr. 219.00
    1 * 1 ROU
       SFr. 219.00
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.