ESD safety shoes, lace-Up, S3, jo_Rapid Low, Jori
Supplier: ELTEN
Certificates
About this item
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
- Padded tongue
- Rubber/PU sole jo_ATHLETIC
- Rubber scuff cap
- Composite toe cap
- Metal- and leather-free
- Available in multiple colour options
Hydrophobised microfibre construction with fabric lining for natural ventilation. Full-length inlay ESD sole. Non-metallic, penetration-resistant midsole.
Certifications: EN ISO 20345 S3 HRO/SRC, form A.
Specifications
- Catalog No:
- ELTN12031/46
- ELTN12031/47
- ELTN12031/48
- ELTN12021/36
- ELTN12021/37
- ELTN12021/38
- ELTN12031/43
- ELTN12031/44
- ELTN12031/45
- ELTN12021/39
- ELTN12021/40
- ELTN12031/36
- ELTN12031/37
- ELTN12021/41
- ELTN12031/38
- ELTN12021/42
- ELTN12031/39
- ELTN12021/43
- ELTN12031/40
- ELTN12021/44
- ELTN12031/41
- ELTN12021/45
- ELTN12031/42
- ELTN12021/46
- ELTN12021/47
- ELTN12021/48
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001