ContactUsLinkComponent
Grade 520 a ½ qualitative filter papers, Whatman™
undefined
Grade 520 a ½ qualitative filter papers, Whatman™
  • Catalog No:
  • WHAT10331494
  • WHAT10331456
  • 515-8066
  • Grade:
  • 520 a ½
  • 520 a ½
  • 520 a ½
  • Weight:
  • 90 g/m²
  • 90 g/m²
  • 90 g/m²
  • Thickness:
  • 300 µm
  • 300 µm
  • 300 µm
  • Particle retention:
  • 15 - 18 µm
  • 15 - 18 µm
  • 15 - 18 µm
  • Filtration speed:
  • 17,5 s (Herzberg)
  • 17,5 s (Herzberg)
  • 17,5 s (Herzberg)
Frequently Bought Together