Magnesia, rods , Ø×H 2×140 mm, for the phosphorus salt pearls
Katalognr. 84109.0100
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic tube
- Pk:100
- Seq:0100
Specifications
Specification Test Results
- Length:about 14 cm
- Diameter:about 2 mm
- Flame test:Passes test
- Pearl test:Passes test
Specification Test Results
About this item
:
Storage Temperature:Ambient
:1309-48-4