Order Entry
Switzerland
ContactUsLinkComponent
Sodium chloride solution 5M DNase-, RNase-, protease-free, Corning®
Sodium chloride solution 5M DNase-, RNase-, protease-free, Corning®
Catalog # MDTC46-032-CV
Supplier:  Corning
CAS Number:  
undefined
Sodium chloride solution 5M DNase-, RNase-, protease-free, Corning®
Catalog # MDTC46-032-CV
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    6
  • Seq:
    2745
  • Pk Info:
    6x 500 ml

Specifications

About this item

 :
 :58.44 g/mol
Storage Temperature:Ambient
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477