Order Entry
Switzerland
ContactUsLinkComponent
Miller's LB broth, Corning®
Miller's LB broth, Corning®
Catalog # MDTC46-050-CM
Supplier:  Corning
CAS Number:  
undefined
Miller's LB broth, Corning®
Catalog # MDTC46-050-CM
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    6
  • Description:
    Miller's LB broth, 1 L

Specifications

About this item

Miller's LB broth is based on the original Lysogeny Broth (LB) media formulation developed by Luria and Bertani, which was designed for the rapid propagation of recombinant E. coli strains.

  • 20-fold concentration sodium chloride compared to the original formulation
  • Widely used for the growth and maintenance of cells
  • Work with plasmid DNA and recombinant protein production
Delivery: Contents: Casein peptone (10 g/L), UF Yeast Powder (5 g/L), NaCl (10 g/L)