Orders LinkContactUsLinkComponent
Pipette tips
Pipette tips
Pipette tips
Pipette tips
These universal fit pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips reduce sample retention.
Product Family Options
  • Tip style
    Volume
    Tip colour
  • QuickRack
    1 - 200 µl
    Yellow
    1 * 960 ST
    1 * 960 ST - SFr. 80.90
    SFr. 80.90
    1 * 960 ST
       SFr. 80.90
  • Extended length
    1 - 300 µl
    Natural
    1 * 960 ST
    1 * 960 ST - SFr. 126.00
    SFr. 126.00
    1 * 960 ST
       SFr. 126.00
  • QuickRack, extended length
    50 - 1250 µl
    Natural
    1 * 768 ST
    1 * 768 ST