ContactUsLinkComponent
Apollo I liquid viewer, with magnification
Apollo I liquid viewer, with magnification
Catalog # ADLM6031301
undefined
Apollo I liquid viewer, with magnification
Catalog # ADLM6031301
Specifications
  • Description:
    Liquid viewer, Apollo I
  • Pk:
    1
Specifications
About this item
Designed for pharmaceutical and laboratory environments, the Apollo I liquid viewer has a polarised inspection light for the detection of particulate matter in transparent liquids contained in ampoules, vials and bottles.
  • Simple detection of unwanted particulate contamination in injections and intravenous infusions
  • Hygienic stainless steel construction
  • LED light source for durable bulb life
  • Intuitive assembly; get started straight away
  • Adjustable height and angle reduce operator fatigue
  • With helpful 1,7× magnifier
Adelphi Manufacturing also offer the Apollo II Liquid Viewer for transparent glass containers, and the Apollo II High Intensity Liquid Viewer for coloured glass containers.
User manual is in English, with additional languages also available upon request.
Certifications: Adelphi Manufacturing’s ISO 9001:2015 certification guarantees the high quality finish of this product. Supporting IQ/OQ documentation can be supplied.
Delivery: The Apollo I includes a storage case for safety and transport.
Packaging: Packaged in sustainably sourced foam, made from polyethylene resin which is derived from renewable sugarcane-based feedstock.