Order Entry
Switzerland
ContactUsLinkComponent
Castroviejo spring scissors, sharply curved up
Castroviejo spring scissors, sharply curved up
Catalog # FSCI15017-10
Supplier:  FINE SCIENCE TOOLS
undefined
Castroviejo spring scissors, sharply curved up
Catalog # FSCI15017-10
Supplier:  FINE SCIENCE TOOLS

Specifications

  • Type:
    Sharp, curved tips
  • Length:
    100 mm
  • Pk:
    1
  • Tip size:
    0,255 mm
  • Blade length:
    1 mm

Specifications

About this item

These scissors are made of stainless steel.