Orders LinkContactUsLinkComponent
Vannas spring scissors, microserrated
Vannas spring scissors, microserrated
Marketsource
Supplier:   FINE SCIENCE TOOLS
Vannas spring scissors, microserrated
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Vannas spring scissors, microserrated
Vannas spring scissors, microserrated
3 Options Available Below
Product Details & Documents
These stainless steel scissors are microserrated to minimise slippage.
Product Family Options
Product Information
  • Type
    Length
    Blade length
    Tip size
    Serrated
    Availability
    Price
  • Sharp, straight tips
    85 mm
    4 mm
    0,1 mm
    +
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 241.39
    SFr. 241.39
    1 * 1 ST
       SFr. 241.39
    Specifications:
    Marketsource
    Marketsource
  • Sharp, curved tips
    85 mm
    4 mm
    0,1 mm
    +
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 245.97
    SFr. 245.97
    1 * 1 ST
       SFr. 245.97
    Specifications:
    Marketsource
    Marketsource
  • Sharp, straight tips
    90 mm
    5 mm
    0,1 mm
    +
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 236.81
    SFr. 236.81
    1 * 1 ST
       SFr. 236.81
    Specifications:
    Marketsource
    Marketsource