Order Entry
Switzerland
ContactUsLinkComponent
2-Phenoxyethanol ≥99.0%, GPR RECTAPUR®
Irritant
2-Phenoxyethanol ≥99.0%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
2-Phenoxyethanol ≥99.0%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (GC, area%):
    min. 99,0%
  • Identification (IR):
    passes test
  • Density (20/4):
    1,105 - 1,110
  • Peroxides (as H₂O₂):
    max. 50 ppm

Specification Test Results

Frequently Bought Together