Anti-Helios Armenian hamster Monoclonal Antibody [clone: 22F6] (Alexa Fluor® 700)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG137242
- BLEG137241
- Antigen symbol:
- Helios
- Helios
- Antigen synonyms:
- IKAROS family zinc finger 2|ZNF1A2|ANF1A2|IKZF2|ZNFN1A2
- IKAROS family zinc finger 2|ZNF1A2|ANF1A2|IKZF2|ZNFN1A2
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 700
- Alexa Fluor® 700
- Clone:
- 22F6
- 22F6
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Armenian hamster
- Armenian hamster
- Isotype:
- IgG
- IgG
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Helios peptide (aa 51-107)
- Helios peptide (aa 51-107)
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 700 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 700 under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA)
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA)
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- ICFC
- ICFC
Specifications
Frequently Bought Together