Anti-IL-10 Rat Monoclonal Antibody [clone: JES5-16E3] (Ultra-LEAF™)
Catalog # BLEG505037
Supplier: Biolegend
Specifications
- Antigen symbol:IL-10
- Clonality:Monoclonal
- Clone:JES5-16E3
- Conjugation:Ultra-LEAF™
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Neutralisation:Yes
- Antigen synonyms:Cytokine synthesis inhibitory factor (CSIF)|Interleukin-10|B cell derived T cell growth factor (B-TCGF)
- Storage buffer:0.2 µm filtered in phosphate-buffered solution, pH 7.2, containing no preservative. Endotoxin level is <0.01 EU/µg of the protein (<0.001 ng/µg of the protein) as determined by the LAL test.
- Storage temperature:2…8 °C
- Immunogen:E. coli expressed, recombinant mouse IL-10
- Tested applications:ELISA Capture, CyTOF®, ELISPOT Capture, Neut, ICFC, IHC
- Purification:The Ultra-LEAF™ (Low Endotoxin, Azide-Free) antibody was purified by affinity chromatography.
- Size:100 µg
- Pk:100 µG