Anti-CD49b Mouse Monoclonal Antibody [clone: P1E6-C5] (PE (Phycoerythrin)/Cy7®)
Catalog # BLEG359314
Supplier: Biolegend
Specifications
- Antigen symbol:CD49b
- Clonality:Monoclonal
- Clone:P1E6-C5
- Conjugation:PE (Phycoerythrin)/Cy7®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen synonyms:VLA-2 α chain|Integrin α2 chain|α2 integrin
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:2…8 °C
- Immunogen:HT1080 cells
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with PE/Cyanine7 under optimal conditions.
- Size:100 tests
- Pk:100 Tests