Anti-CD4 Mouse Monoclonal Antibody [clone: SK3] (PE (Phycoerythrin)/Cy5®)
Catalog # BLEG344654
Supplier: Biolegend
Specifications
- Antigen symbol:CD4
- Clonality:Monoclonal
- Clone:SK3
- Conjugation:PE (Phycoerythrin)-Cy5®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen synonyms:Leu3a|T4
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:2…8 °C
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with PE/Cyanine5 under optimal conditions.
- Size:100 tests
- Pk:100 Tests