Anti-IFN-gamma Mouse Monoclonal Antibody [clone: NIB42] (Ultra-LEAF™)
Catalog # BLEG502406
Supplier: Biolegend
Specifications
- Antigen symbol:IFN-gamma
- Clonality:Monoclonal
- Clone:NIB42
- Conjugation:Ultra-LEAF™
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Neutralisation:Yes
- Antigen synonyms:Type II interferon|T cell interferon|Macrophage-activating factor (MAF)|IFN-gamma|IFN-g|Interferon-γ|Immune interferon
- Storage buffer:0.2 µm filtered in phosphate-buffered solution, pH 7.2, containing no preservative. Endotoxin level is <0.01 EU/µg of the protein (<0.001 ng/µg of the protein) as determined by the LAL test.
- Storage temperature:2…8 °C
- Immunogen:E. coli expressed, recombinant human IFN-γ
- Tested applications:ELISA Capture, ELISPOT Capture, Neut
- Purification:The Ultra-LEAF™ (Low Endotoxin, Azide-Free) antibody was purified by affinity chromatography.
- Size:1 mg
- Pk:1 mg