Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD218a (IL-18Ralpha) Mouse Monoclonal Antibody [clone: H44] (APC (Allophycocyanin))
Anti-CD218a (IL-18Ralpha) Mouse Monoclonal Antibody [clone: H44] (APC (Allophycocyanin))
Catalog # BLEG313814
Anbieter:  Biolegend
Anti-CD218a (IL-18Ralpha) Mouse Monoclonal Antibody [clone: H44] (APC (Allophycocyanin))
Catalog # BLEG313814
Anbieter:  Biolegend

Technische Daten

  • Antigen symbol:
    CD218a (IL-18Ralpha)
  • Clonality:
    Monoclonal
  • Clone:
    H44
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Baboon,
    African green monkey,
    Cynomolgus monkey,
    Rhesus monkey,
    Human
  • Antigen synonyms:
    IL-18 receptor alpha chain|CDw218a|IL-18 receptor 1|IL-18 receptor related protein
  • Storage buffer:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Storage temperature:
    2…8 °C
  • Immunogen:
    Human NK cell line NK0 constitutively expressing IL-18 receptors
  • Tested applications:
    FC
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions.
  • Size:
    100 tests
  • Pk:
    100 Tests

Technische Daten

Über diesen Artikel