Anti-CD1c Mouse Monoclonal Antibody [clone: L161] (Brilliant Violet® 785)
Catalog # BLEG331544
Supplier: Biolegend
Specifications
- Antigen symbol:CD1c
- Clonality:Monoclonal
- Clone:L161
- Conjugation:Brilliant Violet® 785
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Baboon,African green monkey,Cynomolgus monkey,Rhesus monkey,Human
- Antigen synonyms:BDCA-1|M241|R7
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:2…8 °C
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 785™ under optimal conditions.
- Size:100 tests
- Pk:100 Tests