Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (Brilliant Violet® 785)
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (Brilliant Violet® 785)
Catalog # BLEG300234
Supplier:  Biolegend
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (Brilliant Violet® 785)
Catalog # BLEG300234
Supplier:  Biolegend

Specifications

  • Antigen symbol:
    CD2
  • Clonality:
    Monoclonal
  • Clone:
    RPA-2.10
  • Conjugation:
    Brilliant Violet® 785
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Baboon,
    Capuchin monkey,
    Pig,
    African green monkey,
    Cynomolgus monkey,
    Rhesus monkey,
    Pigtailed macaque,
    Chimpanzee,
    Human
  • Antigen synonyms:
    T11|LFA-2|SRBC-R
  • Storage buffer:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Storage temperature:
    2…8 °C
  • Tested applications:
    FC
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 785™ under optimal conditions.
  • Size:
    100 tests
  • Pk:
    100 Tests

Specifications

About this item