Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (Brilliant Violet® 711)
BLEG300232
- Antigen symbol:CD2
- Clonality:Monoclonal
- Clone:RPA-2.10
- Conjugation:Brilliant Violet® 711
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Baboon,Capuchin monkey,Pig,African green monkey,Cynomolgus monkey,Rhesus monkey,Pigtailed macaque,Chimpanzee,Human
- Antigen synonyms:T11|LFA-2|SRBC-R
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:2…8 °C
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711™ under optimal conditions.
- Size:100 tests
- Pk:100 Tests