Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-CD14 Mouse Monoclonal Antibody [clone: 63D3] (Brilliant Violet® 750)
 :  Biolegend
 
Anti-CD14 Mouse Monoclonal Antibody [clone: 63D3] (Brilliant Violet® 750)
 :  Biolegend

 

  • Catalog No:
  • BLEG367136
  • BLEG367135
  • Antigen symbol:
  • CD14
  • CD14
  • Antigen synonyms:
  • Monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein|LPS receptor
  • Monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein|LPS receptor
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Brilliant Violet® 750
  • Brilliant Violet® 750
  • Clone:
  • 63D3
  • 63D3
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Purified human peripheral blood monocytes.
  • Purified human peripheral blood monocytes.
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 750™ under optimal conditions.
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 750™ under optimal conditions.
  • Storage buffer:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Storage temperature:
  • 2…8 °C
  • 2…8 °C
  • Tested applications:
  • FC
  • FC

 

Frequently Bought Together