Anti-MU/HU GL7 Antigen (T/B Cell Act. Marker) Rat Monoclonal Antibody [clone: GL7] (PE (Phycoerythrin)/Cy7®)
Catalog # BLEG144620
Supplier: Biolegend
Specifications
- Antigen symbol:MU/HU GL7 Antigen (T/B Cell Act. Marker)
- Clonality:Monoclonal
- Clone:GL7
- Conjugation:PE (Phycoerythrin)/Cy7®
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgM kappa
- Reactivity:Human,Mouse
- Antigen synonyms:Ly77|T and B cell activation marker
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:2…8 °C
- Concentration:0.2 mg/ml
- Immunogen:LPS activated DBA/J mouse B cells
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with PE/Cyanine7 under optimal conditions.
- Size:100 µg
- Pk:100 µG