Anti-IL-4 Rat Monoclonal Antibody [clone: MP4-25D2] (Ultra-LEAF™)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG500839
- BLEG500838
- Antigen symbol:
- IL-4
- IL-4
- Antigen synonyms:
- MCGF-2|IaIF|HCGF|TCGF-2|MFF|BSF-1|Interleukin-4
- MCGF-2|IaIF|HCGF|TCGF-2|MFF|BSF-1|Interleukin-4
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Ultra-LEAF™
- Ultra-LEAF™
- Clone:
- MP4-25D2
- MP4-25D2
- Reactivity:
- Pig, Rhesus monkey, Human
- Pig, Rhesus monkey, Human
- Host:
- Rat
- Rat
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Immunogen:
- CHO-expressed, recombinant human IL-4
- CHO-expressed, recombinant human IL-4
- Purification:
- The Ultra-LEAF™ (Low Endotoxin, Azide-Free) antibody was purified by affinity chromatography.
- The Ultra-LEAF™ (Low Endotoxin, Azide-Free) antibody was purified by affinity chromatography.
- Storage buffer:
- 0.2 µm filtered in phosphate-buffered solution, pH 7.2, containing no preservative. Endotoxin level is <0.01 EU/µg of the protein (<0.001 ng/µg of the protein) as determined by the LAL test.
- 0.2 µm filtered in phosphate-buffered solution, pH 7.2, containing no preservative. Endotoxin level is <0.01 EU/µg of the protein (<0.001 ng/µg of the protein) as determined by the LAL test.
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- ELISA, CyTOF®, Neut, ICFC, IHC
- ELISA, CyTOF®, Neut, ICFC, IHC
Specifications
Frequently Bought Together