Anti-CD117 (c-kit) Rat Monoclonal Antibody [clone: 2B8] (Alexa Fluor® 700)
N° de catalogue BLEG105846
Fournisseur: Biolegend
Spécifications
- Antigen symbol:CD117 (c-kit)
- Clonality:Monoclonal
- Clone:2B8
- Conjugation:Alexa Fluor® 700
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen synonyms:c-KIT|Stem Cell Factor Receptor (SCFR)
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:2…8 °C
- Concentration:0.5 mg/ml
- Immunogen:Mouse bone marrow mast cells
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 700 under optimal conditions.
- Size:100 µg
- Pk:100 µG