Anti-TCR alpha/beta Mouse Monoclonal Antibody [clone: IP26] (Brilliant Violet® 785)
BLEG306742
- Antigen symbol:TCR alpha/beta
- Clonality:Monoclonal
- Clone:IP26
- Conjugation:Brilliant Violet® 785
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen synonyms:α/β TCR|TCR α/β
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:2…8 °C
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 785™ under optimal conditions.
- Size:100 tests
- Pk:100 Tests