Anti-CD66b Mouse Monoclonal Antibody [clone: 6/40c] (Alexa Fluor® 594)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG392908
- BLEG392907
- Antigen symbol:
- CD66b
- CD66b
- Antigen synonyms:
- CEACAM8|NCA-95|CGM6|CD67
- CEACAM8|NCA-95|CGM6|CD67
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 594
- Alexa Fluor® 594
- Clone:
- 6/40c
- 6/40c
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant soluble human CEACAM8-Fc produced in HEK293 cells
- Recombinant soluble human CEACAM8-Fc produced in HEK293 cells
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 594 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 594 under optimal conditions.
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- IHC-P
- IHC-P
Specifications
Frequently Bought Together