Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-CD196 (CCR6) Mouse Monoclonal Antibody [clone: G034E3] (TotalSeq™-A0143)
  BLEG353437
 :  Biolegend
Anti-CD196 (CCR6) Mouse Monoclonal Antibody [clone: G034E3] (TotalSeq™-A0143)
  BLEG353437
 :  Biolegend

 

  • Antigen symbol:
    CD196 (CCR6)
  • Clonality:
    Monoclonal
  • Clone:
    G034E3
  • Conjugation:
    TotalSeq™-A0143
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Baboon,
    African green monkey,
    Cynomolgus monkey,
    Rhesus monkey,
    Human
  • Antigen synonyms:
    STRL22|GPRCY4|GPR-CY4|CKR-L3|CKRL3|CMKBR6|CKR6|DRY-6|GPR29|BN-1|DCR2
  • Storage buffer:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 1 mM EDTA.
  • Storage temperature:
    2…8 °C
  • Concentration:
    0.5 mg/ml
  • Immunogen:
    CCR6-transfected cells
  • Tested applications:
    PG
  • Purification:
    The antibody was purified by chromatography and conjugated with TotalSeq™-A oligomer under optimal conditions.
  • Size:
    10 µg
  • Pk:
    10 µG