Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (TotalSeq™-C0072)
Catalog # BLEG300567
Anbieter:  Biolegend
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (TotalSeq™-C0072)
Catalog # BLEG300567
Anbieter:  Biolegend

Technische Daten

  • Antigen symbol:
    CD4
  • Clonality:
    Monoclonal
  • Clone:
    RPA-T4
  • Conjugation:
    TotalSeq™-C0072
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Chimpanzee,
    Human
  • Antigen synonyms:
    T4
  • Storage buffer:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 1 mM EDTA.
  • Storage temperature:
    2…8 °C
  • Concentration:
    0.5 mg/ml
  • Tested applications:
    PG
  • Purification:
    The antibody was purified by chromatography and conjugated with TotalSeq™-C oligomer under optimal conditions.
  • Size:
    10 µg
  • Pk:
    10 µG

Technische Daten

Über diesen Artikel