Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD62L Mouse Monoclonal Antibody [clone: DREG-56] (TotalSeq™-B0147)
Anti-CD62L Mouse Monoclonal Antibody [clone: DREG-56] (TotalSeq™-B0147)
  BLEG304849
 :  Biolegend
Anti-CD62L Mouse Monoclonal Antibody [clone: DREG-56] (TotalSeq™-B0147)
  BLEG304849
 :  Biolegend

 

  • Antigen symbol:
    CD62L
  • Clonality:
    Monoclonal
  • Clone:
    DREG-56
  • Conjugation:
    TotalSeq™-B0147
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Chimpanzee,
    Human,
    Bovine
  • Antigen synonyms:
    LAM-1|Leu-8|LECAM-1|TQ-1|L-selectin
  • Storage buffer:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 1 mM EDTA.
  • Storage temperature:
    2…8 °C
  • Concentration:
    0.5 mg/ml
  • Immunogen:
    Concentrated supernatant from PMA-activated human peripheral blood leukocytes
  • Tested applications:
    PG
  • Purification:
    The antibody was purified by chromatography and conjugated with TotalSeq™-B oligomer under optimal conditions.
  • Size:
    10 µg
  • Pk:
    10 µG