Anti-GARP (LRRC32) Rat Monoclonal Antibody [clone: F011-5] (PE (Phycoerythrin)/Cy7®)
BLEG142910
- Antigen symbol:GARP (LRRC32)
- Clonality:Monoclonal
- Clone:F011-5
- Conjugation:PE (Phycoerythrin)/Cy7®
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen synonyms:LRRC32|Glycoprotein A repetitions predominant|Garpin
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:2…8 °C
- Concentration:0.2 mg/ml
- Immunogen:Mouse GARP (extracellular domain)
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with PE/Cyanine7 under optimal conditions.
- Size:100 µg
- Pk:100 µG