Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-IgG Fc Rat Monoclonal Antibody [clone: M1310G05] (PE (Phycoerythrin)/Cy7®)
  BLEG410722-100TST
 :  Biolegend
Anti-IgG Fc Rat Monoclonal Antibody [clone: M1310G05] (PE (Phycoerythrin)/Cy7®)
  BLEG410722-100TST
 :  Biolegend

 

  • Antigen symbol:
    IgG Fc
  • Clonality:
    Monoclonal
  • Clone:
    M1310G05
  • Conjugation:
    PE (Phycoerythrin)/Cy7®
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Storage buffer:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Storage temperature:
    2…8 °C
  • Immunogen:
    Human Siglec-E-IgG Fc fusion protein.
  • Tested applications:
    FC
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with PE/Cyanine7 under optimal conditions.
  • Size:
    100 tests
  • Pk:
    100 Tests