Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD152 (CTLA-4) Mouse Monoclonal Antibody [clone: L3D10] (APC (Allophycocyanin)/FIRE™750)
Anti-CD152 (CTLA-4) Mouse Monoclonal Antibody [clone: L3D10] (APC (Allophycocyanin)/FIRE™750)
Catalog # BLEG349930
Supplier:  Biolegend
Anti-CD152 (CTLA-4) Mouse Monoclonal Antibody [clone: L3D10] (APC (Allophycocyanin)/FIRE™750)
Catalog # BLEG349930
Supplier:  Biolegend

Specifications

  • Antigen symbol:
    CD152 (CTLA-4)
  • Clonality:
    Monoclonal
  • Clone:
    L3D10
  • Conjugation:
    APC (Allophycocyanin)/FIRE™750
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Antigen synonyms:
    Cytotoxic T-Lymphocyte Antigen 4
  • Storage buffer:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Storage temperature:
    2…8 °C
  • Immunogen:
    Extracellular domain of human CTLA-4 and human IgG1 Fc fusion protein
  • Tested applications:
    FC
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with APC/Fire™ 750 under optimal conditions.
  • Size:
    100 tests
  • Pk:
    100 Tests

Specifications

About this item