Anti-CD294 (CRTH2) Rat Monoclonal Antibody [clone: BM16] (Pacific Blue™)
Catalog # BLEG350129
Supplier: Biolegend
Specifications
- Antigen symbol:CD294 (CRTH2)
- Clonality:Monoclonal
- Clone:BM16
- Conjugation:Pacific Blue™
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Human
- Antigen synonyms:GPR44|Chemoattractant receptor-homologous molecule expressed on Th2 cells|DP2|G-protein coupled receptor 44
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:2…8 °C
- Immunogen:Rat cell line TART/B19-12.10 transfected with human CRTH2
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with Pacific Blue™ under optimal conditions.
- Size:25 tests
- Pk:25 Tests