Anti-CD278 (ICOS) Armenian hamster Monoclonal Antibody [clone: C398.4A] (Brilliant Violet® 650)
Catalog # BLEG313549
Supplier: Biolegend
Specifications
- Antigen symbol:CD278 (ICOS)
- Clonality:Monoclonal
- Clone:C398.4A
- Conjugation:Brilliant Violet® 650
- Flow cytometry:Yes
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Baboon,Pig,African green monkey,Cynomolgus monkey,Rhesus monkey,Human,Rat,Mouse
- Antigen synonyms:H4|Inducible COStimulatory molecule
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:2…8 °C
- Immunogen:Mouse T cell clone D10.G4.1
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 650™ under optimal conditions.
- Size:25 tests
- Pk:25 Tests