Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD80 Mouse Monoclonal Antibody [clone: 2D10] (Brilliant Violet® 711)
Anti-CD80 Mouse Monoclonal Antibody [clone: 2D10] (Brilliant Violet® 711)
Catalog # BLEG305236
Supplier:  Biolegend
Anti-CD80 Mouse Monoclonal Antibody [clone: 2D10] (Brilliant Violet® 711)
Catalog # BLEG305236
Supplier:  Biolegend

Specifications

  • Antigen symbol:
    CD80
  • Clonality:
    Monoclonal
  • Clone:
    2D10
  • Conjugation:
    Brilliant Violet® 711
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Rhesus monkey,
    Human
  • Antigen synonyms:
    B7-1|BB1|B7
  • Storage buffer:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Storage temperature:
    2…8 °C
  • Tested applications:
    FC
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711™ under optimal conditions.
  • Size:
    100 tests
  • Pk:
    100 Tests

Specifications

About this item