Anti-CD14 Mouse Monoclonal Antibody [clone: M5E2] (TotalSeq™-A0081)
Catalog # BLEG301855
Anbieter: Biolegend
Technische Daten
- Antigen symbol:CD14
- Clonality:Monoclonal
- Clone:M5E2
- Conjugation:TotalSeq™-A0081
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Capuchin monkey,Squirrel monkey,African green monkey,Cynomolgus monkey,Common marmoset,Rhesus monkey,Cotton-topped tamarin,Pigtailed macaque,Chimpanzee,Human,Bovine,Dog
- Antigen synonyms:LPS receptor
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 1 mM EDTA.
- Storage temperature:2…8 °C
- Concentration:0.5 mg/ml
- Immunogen:Full-length human CD14 protein
- Tested applications:PG
- Purification:The antibody was purified by chromatography and conjugated with TotalSeq™-A oligomer under optimal conditions.
- Size:10 µg
- Pk:10 µG