Anti-CD86 Mouse Monoclonal Antibody [clone: BU63] (PE (Phycoerythrin)/Cy7®)
Catalog # BLEG374210
Supplier: Biolegend
Specifications
- Antigen symbol:CD86
- Clonality:Monoclonal
- Clone:BU63
- Conjugation:PE (Phycoerythrin)/Cy7®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen synonyms:B70|Ly-58|B7-2
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:2…8 °C
- Immunogen:ARH 77 (B lymphoblastoid cell line).
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with PE/Cyanine7 under optimal conditions.
- Size:100 tests
- Pk:100 Tests