Anti-STAT1 (phospho-Tyr701) Mouse Monoclonal Antibody [clone: A17012A] (PE (Phycoerythrin))
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG666404
- BLEG666403
- Antigen symbol:
- STAT1
- STAT1
- Antigen synonyms:
- Transcription Factor ISGF-3 components p91/p84|Signal Transduced Activator of Transcription 1
- Transcription Factor ISGF-3 components p91/p84|Signal Transduced Activator of Transcription 1
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- A17012A
- A17012A
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Modification:
- phospho-Tyr701
- phospho-Tyr701
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- ICFC
- ICFC
Specifications
Frequently Bought Together