Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD282 (TLR2) Mouse Recombinant Antibody [clone: QA16A01] (Alexa Fluor® 647)
Anti-CD282 (TLR2) Mouse Recombinant Antibody [clone: QA16A01] (Alexa Fluor® 647)
Catalog # BLEG153008
Supplier:  Biolegend
Anti-CD282 (TLR2) Mouse Recombinant Antibody [clone: QA16A01] (Alexa Fluor® 647)
Catalog # BLEG153008
Supplier:  Biolegend

Specifications

  • Antigen symbol:
    CD282 (TLR2)
  • Clonality:
    Recombinant
  • Clone:
    QA16A01
  • Conjugation:
    Alexa Fluor® 647
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Mouse
  • Antigen synonyms:
    Toll-like receptor 2|TLR2|TLR-2|CD282|TIL4
  • Storage buffer:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Storage temperature:
    2…8 °C
  • Concentration:
    0.5 mg/ml
  • Immunogen:
    Recombinant extracellular domain of murine TLR2
  • Tested applications:
    FC
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 647 under optimal conditions.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item