Anti-beta-Amyloid, 17-24 Mouse Monoclonal Antibody [clone: 4G8] (Alexa Fluor® 594)
- Catalog No:
- BLEG800715
- BLEG800716
- Antigen symbol:
- beta-Amyloid, 17-24
- beta-Amyloid, 17-24
- Antigen synonyms:
- APP|CTFgamma|CVAP|protease nexin-II|ABETA|cerebral vascular amyloid peptide|beta-amyloid peptide|APPI|AD1|PN-II|Amyloid Precursor Protein|preA4|ABPP|alzheimer disease amyloid protein|Amyloid beta A4 protein|AAA|PN2|peptidase nexin-II
- APP|CTFgamma|CVAP|protease nexin-II|ABETA|cerebral vascular amyloid peptide|beta-amyloid peptide|APPI|AD1|PN-II|Amyloid Precursor Protein|preA4|ABPP|alzheimer disease amyloid protein|Amyloid beta A4 protein|AAA|PN2|peptidase nexin-II
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 594
- Alexa Fluor® 594
- Clone:
- 4G8
- 4G8
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b kappa
- IgG2b kappa
- ImmunoChemistry:
- Yes
- Yes
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 594 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 594 under optimal conditions.
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- IHC-P
- IHC-P
Frequently Bought Together