Anti-CD209 (DC-SIGN) Mouse Monoclonal Antibody [clone: 9E9A8] (APC (Allophycocyanin)/FIRE™750)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG330115
- BLEG330116
- Antigen symbol:
- CD209 (DC-SIGN)
- CD209 (DC-SIGN)
- Antigen synonyms:
- Dendritic Cell-Specific Intercellular adhesion molecule 3 (ICAM-3)-Grabbing Nonintegrin
- Dendritic Cell-Specific Intercellular adhesion molecule 3 (ICAM-3)-Grabbing Nonintegrin
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)/FIRE™750
- APC (Allophycocyanin)/FIRE™750
- Clone:
- 9E9A8
- 9E9A8
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Extracellular domain of human DC-SIGN
- Extracellular domain of human DC-SIGN
- Purification:
- The antibody was purified by affinity chromatography and conjugated with APC/Fire™ 750 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with APC/Fire™ 750 under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Specifications
Frequently Bought Together