Anti-CD206 (MMR) Mouse Monoclonal Antibody [clone: 15-2] (Brilliant Violet® 711)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG321135
- BLEG321136
- Antigen symbol:
- CD206 (MMR)
- CD206 (MMR)
- Antigen synonyms:
- CD206|MMR (macrophage mannose receptor)|MRC1|MR (mannose receptor)
- CD206|MMR (macrophage mannose receptor)|MRC1|MR (mannose receptor)
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Brilliant Violet® 711
- Brilliant Violet® 711
- Clone:
- 15-2
- 15-2
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Purified human mannose receptor
- Purified human mannose receptor
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711™ under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711™ under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Specifications
Frequently Bought Together